Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Notch-1 Alexa Fluor 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB5267S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB5267S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB5267S100 Supplier R&D Systems Supplier No. FAB5267S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Notch-1 Monoclonal specifically detects Notch-1 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen Notch-1
Applications Flow Cytometry
Classification Monoclonal
Clone N1A
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q01705
Gene Alias EC 2.1.2.11, EC 3.4.21.68, hN1, Notch (Drosophila) homolog 1 (translocation-associated), notch 1Notch homolog 1, translocation-associated (Drosophila), Notch homolog 1, translocation-associated, Notch1, TAN1, TAN1neurogenic locus notch homolog protein 1, Translocation-associated notch protein TAN-1
Gene Symbols NOTCH1
Host Species Mouse
Immunogen Mouse embryonic fibroblast cell line PA317-derived recombinant mouse Notch-1 Accession # Q01705
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4851
Test Specificity Detects mouse Notch-1 intracellular domain in direct ELISAs.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.