Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Notch-2 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB5196R100UG
This item is not returnable.
View return policy
Description
Notch-2 Monoclonal specifically detects Notch-2 in Mouse samples. It is validated for Flow Cytometry.Specifications
Notch-2 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
O35516 | |
NOTCH2 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse Notch-2 Leu26-Val528 (predicted) Accession # O35516 | |
RUO | |
4853 | |
Mouse | |
IgG1 |
Flow Cytometry | |
605724 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
AGS2, hN2, neurogenic locus notch homolog protein 2, Notch (Drosophila) homolog 2, notch 2Notch homolog 2 (Drosophila), Notch homolog 2, Notch2 | |
Rat | |
100 μg | |
Primary | |
Detects mouse Notch-2 in direct ELISAs. In direct ELISAs, 50-100% cross-reactivity with recombinant human (rh) Notch-2 and no cross-reactivity with rhNotch-1, -2, -3, recombinant mouse (rm) Notch-1, rmNotch-3, recombinant rat (rr) Notch2, or rrDLL1 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction