Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Notch-2 Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB5196N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB5196N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB5196N100 Supplier R&D Systems Supplier No. FAB5196N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Notch-2 Monoclonal specifically detects Notch-2 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Notch-2
Applications Flow Cytometry
Classification Monoclonal
Clone 605724
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. O35516
Gene Alias AGS2, hN2, neurogenic locus notch homolog protein 2, Notch (Drosophila) homolog 2, notch 2Notch homolog 2 (Drosophila), Notch homolog 2, Notch2
Gene Symbols NOTCH2
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Notch-2 Leu26-Val528 (predicted) Accession # O35516
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4853
Test Specificity Detects mouse Notch-2 in direct ELISAs. In direct ELISAs, 50-100% cross-reactivity with recombinant human (rh) Notch-2 and no cross-reactivity with rhNotch-1, -2, -3, recombinant mouse (rm) Notch-1, rmNotch-3, recombinant rat (rr) Notch2, or rrDLL1 is observed.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.