Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Notch-3 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
$227.50 - $584.50
Specifications
Antigen | Notch-3 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Blockade of Receptor-ligand Interaction, CyTOF |
Classification | Polyclonal |
Description
Notch-3 Polyclonal specifically detects Notch-3 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Blockade of Receptor-ligand Interaction, CyTOF-ready.Specifications
Notch-3 | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
Polyclonal | |
Goat | |
Mouse | |
CADASIL, CASILneurogenic locus notch homolog protein 3, Notch (Drosophila) homolog 3, notch 3Notch homolog 3 (Drosophila), Notch homolog 3, Notch3 | |
NOTCH3 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Flow Cytometry, Immunohistochemistry, Blockade of Receptor-ligand Interaction, CyTOF | |
Unconjugated | |
RUO | |
Q61982 | |
4854 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Notch-3 Ala40-Glu468 Accession # Q61982 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Notch-3 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human Notch-3 is observed and less than 5% cross-reactivity with recombinant rat (rr) Notch-1 and rrNotch-2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title