Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse NOV/CCN3 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB19762SP
This item is not returnable.
View return policy
Description
NOV/CCN3 Monoclonal specifically detects NOV/CCN3 in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
NOV/CCN3 | |
Monoclonal | |
LYOPH | |
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Q64299 | |
NOV | |
Mouse myeloma cell line NS0-derived recombinant mouse NOV/CCN3 Ser26-Ile354 Accession # Q64299 | |
25 μg | |
Primary | |
Detects mouse NOV/CCN3 in ELISAs. | |
Mouse | |
Purified |
ELISA | |
231226 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CCN family member 3, CCN3, CCN3IGF-binding protein 9, IBP-9, IGFBP-9, IGFBP9novH, Insulin-like growth factor-binding protein 9, nephroblastoma overexpressed gene, Nephroblastoma-overexpressed gene protein homolog, NOVH, protein NOV homolog | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4856 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction