Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse NOV/CCN3 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB1976
This item is not returnable.
View return policy
Description
NOV/CCN3 Monoclonal specifically detects NOV/CCN3 in Mouse samples. It is validated for Western Blot.Specifications
| NOV/CCN3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q64299 | |
| NOV | |
| Mouse myeloma cell line NS0-derived recombinant mouse NOV/CCN3 Ser26-Ile354 Accession # Q64299 | |
| 500 μg | |
| Primary | |
| Detects mouse NOV/CCN3 in direct ELISAs and Western blots. No cross-reactivity with recombinant human (rh) CTGF, rhNOV/CCN3, or recombinant mouse WISP-1 is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 231216 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CCN family member 3, CCN3, CCN3IGF-binding protein 9, IBP-9, IGFBP-9, IGFBP9novH, Insulin-like growth factor-binding protein 9, nephroblastoma overexpressed gene, Nephroblastoma-overexpressed gene protein homolog, NOVH, protein NOV homolog | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4856 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction