Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse NT-4 ELISA Kit (Colorimetric)
SDP

Catalog No. NBP276769 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP276769 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP276769 Supplier Novus Biologicals™ Supplier No. NBP276769
Add to Cart
Add to Cart

Mouse NT-4 ELISA Kit (Colorimetric) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse NT-4 ELISA Kit (Colorimetric) measures mouse NT-4 in serum, plasma and other biological fluids.

Specifications

Assay Range 15.625 to 1000 pg/mL
Assay Sensitivity 9.375 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Biological Fluids, Plasma, Serum
For Use With (Application) Western Blot, ELISA
Gene ID (Entrez) 4909
Gene Symbol NTF4
Host Species Mouse
Quantity 1 Kit
Storage Requirements Storage of components varies. See protocol for specific instructions.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.