Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse NTB-A/SLAMF6 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3986
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3986 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3986 Supplier R&D Systems Supplier No. AF3986
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

NTB-A/SLAMF6/CD352 Polyclonal specifically detects NTB-A/SLAMF6/CD352 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.

Specifications

Antigen NTB-A/SLAMF6/CD352
Applications Western Blot, Flow Cytometry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Gene Accession No. Q9ET39
Gene Alias Activating NK receptor, CD352, CD352 antigen, KALIFLJ50657, Ly108, NK-T-B-antigen, NTBA, NTBA receptor, NTB-AMGC104953, NTBAT- and B-cell antigen, SF2000, SLAM family member 6, SLAMF6
Gene Symbols SLAMF6
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse NTB-A/SLAMF6 Glu31-Asn239 Accession # Q9ET39
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114836
Test Specificity Detects mouse NTB-A/SLAMF6 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.