Learn More
Description
Includes
Mouse OSM Capture Antibody; Mouse OSM Detection Antibody; Mouse OSM Standard; Streptavidin-HRP
Specifications
Specifications
| Product Type | Solid Phase Sandwich ELISA |
| Quantity | 5 Plates |
| Species | Mouse |
| Conjugate | Biotin |
| For Use With (Application) | ELISA |
| Product Line | R&D Systems |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Research Area | Immunology |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
