Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Opticin Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3547
This item is not returnable.
View return policy
Description
Opticin Polyclonal specifically detects Opticin in Mouse samples. It is validated for Western Blot.Specifications
Opticin | |
Polyclonal | |
Unconjugated | |
Q920A0 | |
OPTC | |
Mouse myeloma cell line NS0-derived recombinant mouse Opticin Ala20-Thr328 (Tyr168Asp) Accession # Q920A0 | |
100 μg | |
Primary | |
Detects mouse Opticin in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant human Opticin is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
oculoglycan, Oculogylcan, OPT, OPTC, opticin | |
Goat | |
Affinity Purified | |
RUO | |
26254 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction