Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Osteoactivin/GPNMB Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2330
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2330 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2330 Supplier R&D Systems Supplier No. AF2330
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Osteoactivin/GPNMB Polyclonal specifically detects Osteoactivin/GPNMB in Mouse samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen Osteoactivin/GPNMB
Applications Western Blot, Neutralization, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Neutralization 0.2-0.8 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL
Gene Accession No. Q8BVA0
Gene Alias DC-HIL, glycoprotein (transmembrane) nmb, glycoprotein NMB, GPNMB, HGFIN, HGFINglycoprotein nmb-like protein, NMBosteoactivin, Transmembrane glycoprotein HGFIN, transmembrane glycoprotein NMB
Gene Symbols GPNMB
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Osteoactivin/GPNMB Lys23-Asn502 Accession # Q8BVA0
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10457
Test Specificity Detects mouse Osteoactivin/GPNMB in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity with recombinant human (rh) Osteoactivin, rhSyndecan-4, and recombinant mouse Syndecan-4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.