Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse Osteoprotegerin/TNFRSF11B ELISA Kit (Colorimetric)
SDP

Catalog No. NB601001KT Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB601001KT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB601001KT Supplier Novus Biologicals™ Supplier No. NBP2601001KIT

Colorimetric ELISA Kit useful in study of immunity, chemokine, chemotaxis

The Mouse Osteoprotegerin/TNFRSF11B ELISA Kit (Colorimetric) measures mouse Osteoprotegerin/TNFRSF11B in serum, plasma, tissue homogenates.

Specifications

Assay Range 15.6 to 1000pg/mL
Assay Sensitivity 3.9pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Plasma, Serum, Tissue Homogenates
Specificity Detection of mouse OPG
Target Species Mouse
For Use With (Application) ELISA
Gene ID (Entrez) 4982
Gene Symbol TNFRSF11B
Kit Contents Assay plate (12 x 8 coated Microwells), Standard (Freeze dried), Biotin-antibody (100X concentrate), HRP-avidin (100X concentrate), Biotin-antibody Diluents, HRP-avidin Diluents, Sample Diluents, Wash Buffer (25X concentrate), TMB Substrate, Stop Solution, Adhesive Strip (For 96 wells), Instruction manual
Quantity 1 Kit
Regulatory Status RUO
Research Discipline Cell Biology
Sample Volume 50 to 100 μL
Storage Requirements Store unopened product at 2° to 8°C Protect from light. Do not use past expiration date.
Synonym OCIFMGC29565, OPGtumor necrosis factor receptor superfamily member 11B, Osteoclastogenesis inhibitory factor, Osteoprotegerin, TR1, tumor necrosis factor receptor superfamily, member 11b
Target Osteoprotegerin/TNFRSF11B
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.