Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse OX40 Ligand/TNFSF4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1236100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1236100 100 μg
MAB1236SP 25 μg
MAB1236500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB1236100 Supplier R&D Systems Supplier No. MAB1236100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 3 publications

OX40 Ligand/TNFSF4 Monoclonal specifically detects OX40 Ligand/TNFSF4 in Mouse samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen OX40 Ligand/TNFSF4
Applications Western Blot, Neutralization
Classification Monoclonal
Clone 182601
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.01-0.05 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P43488
Gene Alias CD134 ligand, CD134L, CD252, CD252 antigen, Glycoprotein Gp34, gp34, OX40 antigen ligand, OX40 ligand, OX40L, OX-40L, OX4OL, TAX transcriptionally-activated glycoprotein 1, tax-transcriptionally activated glycoprotein 1 (34kD), tax-transcriptionally activated glycoprotein 1, 34kD, TNFSF4, tumor necrosis factor (ligand) superfamily, member 4, tumor necrosis factor ligand superfamily member 4, TXGP1
Gene Symbols TNFSF4
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse OX40 Ligand/TNFSF4 Gln49-Leu198 Accession # P43488
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7292
Test Specificity Detects mouse OX40 Ligand/TNFSF4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) APRIL, rhGITR Ligand, rhLIGHT, rhLT alpha 1/beta 2, rhLT alpha 2/beta 1, recombinant cotton rat TNF-alpha , recombinant mouse (rm) TNF-alpha , rmTRAIL, rhTRANCE, rmTRANCE, rmTWEAK, or rhVEGI is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.