Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse OX40 Ligand/TNFSF4 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $655.00
Specifications
Antigen | OX40 Ligand/TNFSF4 |
---|---|
Clone | 182609 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB12362SP
![]() |
R&D Systems
MAB12362SP |
25 μg |
Each for $200.50
|
|
|||||
MAB12362
![]() |
R&D Systems
MAB12362 |
500 μg |
Each for $655.00
|
|
|||||
Description
OX40 Ligand/TNFSF4 Monoclonal specifically detects OX40 Ligand/TNFSF4 in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
OX40 Ligand/TNFSF4 | |
LYOPH | |
Western Blot, ELISA | |
Unconjugated | |
Rat | |
Mouse | |
P43488 | |
7292 | |
Mouse myeloma cell line NS0-derived recombinant mouse OX40 Ligand/TNFSF4 Gln49-Leu198 Accession # P43488 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse OX40 Ligand/TNFSF4 in ELISAs and Western blots. In sandwich immunoassays, no cross-reactivity or interference with recombinant mouse (rm) OX40, recombinant human (rh) OX40 Ligand, rmFas Ligand, rmLIGHT, or rhLIGHT is observed. In Western blots, 100% cross-reactivity with rmTNF-alpha is observed, 15-30% with rmTRANCE and rmTRAIL is observed, and no cross-reactivity with rmBAFF, rmEDA, rmGITR Ligand, rmLIGHT, rmTWEAK, rmFas Ligand, rhAPRIL, or rhOX40 Ligand is observed. |
182609 | |
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD134 ligand, CD134L, CD252, CD252 antigen, Glycoprotein Gp34, gp34, OX40 antigen ligand, OX40 ligand, OX40L, OX-40L, OX4OL, TAX transcriptionally-activated glycoprotein 1, tax-transcriptionally activated glycoprotein 1 (34kD), tax-transcriptionally activated glycoprotein 1, 34kD, TNFSF4, tumor necrosis factor (ligand) superfamily, member 4, tumor necrosis factor ligand superfamily member 4, TXGP1 | |
TNFSF4 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title