Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse OX40 Ligand/TNFSF4 PE-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems FAB1236P
This item is not returnable.
View return policy
Description
OX40 Ligand/TNFSF4 Polyclonal specifically detects OX40 Ligand/TNFSF4 in Mouse samples. It is validated for Flow Cytometry.Specifications
OX40 Ligand/TNFSF4 | |
Polyclonal | |
Flow Cytometry 10 uL/10^6 cells | |
CD134 ligand, CD134L, CD252, CD252 antigen, Glycoprotein Gp34, gp34, OX40 antigen ligand, OX40 ligand, OX40L, OX-40L, OX4OL, TAX transcriptionally-activated glycoprotein 1, tax-transcriptionally activated glycoprotein 1 (34kD), tax-transcriptionally activated glycoprotein 1, 34kD, TNFSF4, tumor necrosis factor (ligand) superfamily, member 4, tumor necrosis factor ligand superfamily member 4, TXGP1 | |
Goat | |
Affinity Purified | |
RUO | |
7292 | |
Mouse | |
IgG |
Flow Cytometry | |
R-PE | |
P43488 | |
TNFSF4 | |
Mouse myeloma cell line NS0-derived recombinant mouse OX40 Ligand/TNFSF4 Gln49-Leu198 Accession # P43488 | |
100 Tests | |
Primary | |
Detects mouse OX40 Ligand/TNFSF4 in direct ELISAs and Western blots. In Western blots, approximately 5% cross-reactivity with recombinant human (rh) OX40 Ligand, rhLIGHT, recombinant mouse (rm) Fas Ligand and less than 1% cross-reactivity with rmTNF-alpha , rhGITR Ligand, rhTRANCE, and rhVEGI is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction