Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse OX40/TNFRSF4 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF1256
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF1256 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF1256 Supplier R&D Systems Supplier No. BAF1256
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

OX40/TNFRSF4 Polyclonal specifically detects OX40/TNFRSF4 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen OX40/TNFRSF4
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. P47741
Gene Alias ACT-135, ACT35 antigen, ACT35ATC35 antigen, CD134, CD134 antigen, Ly-70, OX40 cell surface antigen, OX40 homologue, OX40L receptor, OX40lymphoid activation antigene ACT35, TAX transcriptionally-activated glycoprotein 1 receptor, tax-transcriptionally activated glycoprotein 1 receptor, TNFRSF4, tumor necrosis factor receptor superfamily member 4, tumor necrosis factor receptor superfamily, member 4, Txgp1, TXGP1L, TXGP1LOX40 antigen
Gene Symbols TNFRSF4
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse OX40/TNFRSF4 Val20-Pro211 Accession # P47741
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7293
Test Specificity Detects mouse OX40/TNFRSF4 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant mouse (rm) 4-1BB, rmCD27, rmCD30, rmCD40, rmDR3, rmEDAR, rmFas, rmGITR, rmOPG, rmRANK, rmTNF RI, and rmTNF RII is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.