Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Mouse Oxidized LDL ELISA Kit (Colorimetric)

Click to view available options
Quantity:
1 Kit
Specifications
Specifications
Assay Range | 0.31-20 ng/mL |
Assay Sensitivity | 0.016 ng/mL |
Product Type | ELISA Kit (Colorimetric) |
Sample Type | Biological Fluids, Plasma, Serum |
For Use With (Application) | ELISA, Sandwich ELISA |
Quantity | 1 Kit |
Storage Requirements | Storage of components varies. See protocol for specific instructions. |
Synonym | Oxidized LDL, Oxidized Low Density Lipoprotein, oxLDL |
Target | Mouse |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction