Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse P-Selectin/CD62P Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB737SP
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB737SP 25 μg
MAB737 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB737SP Supplier R&D Systems Supplier No. MAB737SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 2 publications

P-Selectin/CD62P Monoclonal specifically detects P-Selectin/CD62P in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen P-Selectin/CD62P
Applications Western Blot, ELISA
Classification Monoclonal
Clone 127933
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q01102
Gene Alias CD62P, CD62P antigen, FLJ45155, GMP140, GRMP, PADGEM, PADGEMantigen CD62), PSEL, selectin P (granule membrane protein 140kDa, antigen CD62), SELP
Gene Symbols SELP
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse P-Selectin/CD62P Trp42-Ala709 Accession # Q01102
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6403
Test Specificity Detects mouse P-Selectin/CD62P in ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse (rm) L-Selectin, rmE-Selectin, or recombinant human P-Selectin is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.