Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse PCK1 ELISA Kit (Colorimetric)
SDP

Catalog No. NB016768 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB016768 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB016768 Supplier Novus Biologicals™ Supplier No. NBP281209
Add to Cart
Add to Cart

This kit recognizes Mouse PCK1 in samples.

No significant cross-reactivity or interference between Mouse PCK1 and analogues was observed.

Specifications

Assay Range 0.16 to 10 ng/mL
Assay Sensitivity 0.09 ng/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 5105
Gene Symbol PCK1
Host Species Mouse
Quantity 1 Kit
Research Discipline Lipid and Metabolism
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target PCK1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.