Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse PD-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7738
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7738 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7738 Supplier R&D Systems Supplier No. MAB7738
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

PD-1 Monoclonal specifically detects PD-1 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen PD-1
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 766104
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q02242
Gene Alias CD279, CD279 antigen, hPD-1, PD1, PD1hPD-l, PDCD1, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2
Gene Symbols PDCD1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse PD-1 Leu25-Gln167 Accession # Q02242
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5133
Test Specificity Detects mouse PD-1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human PD-1 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.