Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse PD-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB77382100
This item is not returnable.
View return policy
Description
PD-1 Monoclonal antibody specifically detects PD-1 in Mouse samples. It is validated for NeutralizationSpecifications
PD-1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
CD279, CD279 antigen, hPD-1, PD1, PD1hPD-l, PDCD1, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
Mouse myeloma cell line NS0-derived recombinant mouse PD-1, Met1-Gln167, Accession # Q02242 | |
100 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
IgG2a |
Neutralization | |
996209 | |
Neutralization | |
Q02242 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5133 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction