Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse PD-L1/B7-H1 Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. MAB9078SP Shop All Novus Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9078SP 25 μg
MAB9078 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9078SP Supplier R&D Systems Supplier No. MAB9078SP
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rat Monoclonal Antibody

PD-L1/B7-H1 Monoclonal specifically detects PD-L1/B7-H1 in Mouse samples. It is validated for CyTOF∣Flow Cytometry∣Immunohistochemistry.

Specifications

Antigen PD-L1/B7-H1
Applications CyTOF, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 929903
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9EP73
Gene Alias Avelumab, B7-H, B7H1, B7-H1, B7H1PDCD1L1, CD274, CD274 antigenMGC142294, CD274 molecule, PDCD1L1, PDCD1LG1, PDCD1LG1MGC142296, PDL1, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1
Gene Symbols Cd274
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse PD-L1/B7-H1 Met1-Thr239 Accession # Q9EP73
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29126
Test Specificity Detects mouse PD-L1/B7-H1 in direct ELISAs. Stains mouse PD-L1/B7-H1 transfectants but not irrelevant transfectants in flow cytometry.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Type Antibody
Form Purified
Isotype IgG2a
Product Line R&D Systems
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.