Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse PD-L1/B7-H1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody has been used in 4 publications
$214.00 - $573.00
Specifications
Antigen | PD-L1/B7-H1 |
---|---|
Clone | 2096C |
Dilution | Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB90781SP
![]() |
R&D Systems
MAB90781SP |
25 μg |
Each for $214.00
|
|
|||||
MAB90781100
![]() |
R&D Systems
MAB90781100 |
100 μg |
Each for $573.00
|
|
|||||
Description
PD-L1 Monoclonal specifically detects PD-L1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
PD-L1/B7-H1 | |
Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL | |
Monoclonal | |
Rabbit | |
Mouse | |
Q9EP73 | |
29126 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
2096C | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Avelumab, B7-H, B7H1, B7-H1, B7H1PDCD1L1, CD274, CD274 antigenMGC142294, CD274 molecule, PDCD1L1, PDCD1LG1, PDCD1LG1MGC142296, PDL1, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 | |
Mouse myeloma cell line NS0-derived recombinant mouse PD-L1/B7-H1 Met1-His239 Accession # Q9EP73 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse PD-L1/B7-H1 in direct ELISAs and Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title