Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse PD-L2/B7-DC Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB1022G100UG
This item is not returnable.
View return policy
Description
PD-L2/B7-DC/PDCD1LG2 Monoclonal specifically detects PD-L2/B7-DC/PDCD1LG2 in Mouse samples. It is validated for Flow Cytometry.Specifications
PD-L2/B7-DC/PDCD1LG2 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9WUL5 | |
PDCD1LG2 | |
Mouse myeloma cell line NS0-derived recombinant mouse PD-L2/B7-DC Met1-Arg219 Accession # Q9WUL5 | |
RUO | |
80380 | |
Mouse | |
IgG2a |
Flow Cytometry | |
168633 | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
B7-DC, bA574F11.2, Btdc, Butyrophilin B7-DC, Butyrophilin-like Protein, CD273, CD273 antigen, CD273PD-1 ligand 2, MGC142240, PD-1-ligand 2, PDCD1L2MGC142238, PDCD1LG2, PDL2, PDL2B7DC, PD-L2PDCD1 ligand 2, programmed cell death 1 ligand 2, Programmed death ligand 2 | |
Rat | |
100 μg | |
Primary | |
Detects mouse PD-L2/B7-DC in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant human (rh) PD-L2 is observed and no cross-reactivity with rhB7-1, recombinant mouse (rm) B7-1, rmB7-2, rmB7-H1, rmB7-H2, rmB7-H3, or rmB7-H4 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction