Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse PDGF R alpha Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
$200.50 - $667.00
Specifications
Antigen | PDGFR alpha |
---|---|
Clone | 189208 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
PDGFR alpha Monoclonal specifically detects PDGFR alpha in Mouse samples. It is validated for Western Blot.Specifications
PDGFR alpha | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
P26618 | |
5156 | |
Mouse myeloma cell line NS0-derived recombinant mouse PDGF R alpha Leu25-Glu524 (Asp65Glu, Gly439Ala, Thr440Ala), predicted Accession # P26618 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse PDGF R alpha in direct ELISAs and Western blots. In direct ELISAs and Western blots, 5-20% cross-reactivity with recombinant human PDGF R alpha is observed and no cross-reactivity with recombinant mouse PDGF R beta is observed. |
189208 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR alpha, PDGFR2, PDGFRA, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA | |
PDGFRA | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title