Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Persephin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2479
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2479 100 μg
AF2479SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2479 Supplier R&D Systems Supplier No. AF2479
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Persephin Polyclonal specifically detects Persephin in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Persephin
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. O70300.1
Gene Alias persephin, PSP, PSPN
Gene Symbols PSPN
Host Species Sheep
Immunogen E. coli-derived recombinant mouse Persephin Ala61-Gly156 Accession # O70300.1
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5623
Test Specificity Detects mouse Persephin in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human Persephin is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.