Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse PGLYRP2/PGRP-L Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB4704SP
This item is not returnable.
View return policy
Description
PGLYRP2/PGRP-L Monoclonal specifically detects PGLYRP2/PGRP-L in Mouse samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
PGLYRP2/PGRP-L | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Q8VCS0 | |
PGLYRP2 | |
Mouse myeloma cell line NS0-derived recombinant mouse PGLYRP2/PGRP-L Ser23-Asn530 Accession # Q8VCS0 | |
25 μg | |
Primary | |
Detects mouse PGLYRP2/PGRP-L in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse (rm) PGRP-S and rmPGRP-1 beta is observed. | |
Mouse | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
439728 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.5.1.28, peptidoglycan recognition protein 2tagl-beta, peptidoglycan recognition protein L, PGLYRPL, PGLYRPLHMFT0141, PGRP2, PGRPL, PGRP-L, PGRP-LN-acetylmuramoyl-L-alanine amidase, PGRPLPeptidoglycan recognition protein long, tagL, tagL-alpha, TAGL-like | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
114770 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction