Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse Phospho-Tie-2 DuoSet IC ELISA
SDP

Catalog No. DYC28165 Shop All Novus Biologicals Products
Change view
Click to view available options
Packaging Quantity:
2 Plates
5 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
DYC28165 5 Plates
DYC28162 2 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. DYC28165 Supplier R&D Systems™ Supplier No. DYC28165

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Phospho-Tie-2 DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse Phospho-Tie-2 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated mouse Tie-2.

Specifications

Product Type ELISA Development Kit for Phosphorylated Intracellular Proteins
Quantity 1 Kit
Species Mouse
Conjugate Biotin
For Use With (Application) ELISA
Gene ID (Entrez) 21687
Product Line R&D Systems
Packaging Quantity 5 Plates
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Research Area Signaling
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.