Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse PILR-beta Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | PILR-beta |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
PILR-beta Polyclonal specifically detects PILR-beta in Mouse samples. It is validated for Western Blot.Specifications
PILR-beta | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
activating receptor PILRbeta, Activating receptor PILR-beta, Cell surface receptor FDFACT, cell surface receptor FDFACT1, cell surface receptor FDFACT2, FDFACT, FDFACT1, FDFACT2, paired immunoglobin-like receptor beta, paired immunoglobin-like type 2 receptor beta, paired immunoglobulin-like receptor beta, paired immunoglobulin-like type 2 receptor beta, PILRB, PILRbeta | |
Pilrb1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q2YFS2 | |
29990 | |
Mouse myeloma cell line NS0-derived recombinant mouse PILR-beta Ala28-Gly193 Accession # Q2YFS2 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse PILR-beta in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 20% cross-reactivity with recombinant mouse PILR-alpha is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title