Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse PIR-A/B Alexa Fluor 594-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB5308T100UG
This item is not returnable.
View return policy
Description
PIR-A/B Monoclonal specifically detects PIR-A/B in Mouse samples. It is validated for Flow Cytometry.Specifications
PIR-A/B | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
A2NTJ8 | |
Mouse myeloma cell line NS0-derived recombinant mouse PIR-A1 Gly24-Met650 Accession # A2NTJ8 | |
RUO | |
Detects mouse PIR-A and mouse PIR-B in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Flow Cytometry | |
404127 | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Rat | |
100 μg | |
Primary | |
Mouse | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction