Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse PIR-B Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB2754N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2754N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2754N100 Supplier R&D Systems Supplier No. FAB2754N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

PIR-B Monoclonal specifically detects PIR-B in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen PIR-B
Applications Flow Cytometry
Classification Monoclonal
Clone 326414
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. AAH26937
Gene Alias CD85a, HL9, ILT5, ILT-5, leukocyte immunoglobulin like receptor B3, LILRA6, LILRB3, LIR3, LIR-3, PIRB, PIR-B
Gene Symbols Pirb
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse PIR-B Gly24-Gly635 Accession # AAH26937
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11025
Test Specificity Detects mouse PIR-B in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant mouse PIR-A.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.