Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse PLA2G2A Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF4925
This item is not returnable.
View return policy
Description
PLA2G2A Polyclonal specifically detects PLA2G2A in Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.Specifications
| PLA2G2A | |
| Polyclonal | |
| Unconjugated | |
| NP_001076000 | |
| PLA2G2A | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse PLA2G2A Asn22-Cys146 Accession # NP_001076000 | |
| 100 μg | |
| Primary | |
| Detects mouse PLA2G2A in Western blots. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| EC 3.1.1.4, GIIC sPLA2, Group IIA phospholipase A2, MOM1, Non-pancreatic secretory phospholipase A2, NPS-PLA2, Phosphatidylcholine 2-acylhydrolase 2A, phospholipase A2, group IIA (platelets, synovial fluid), phospholipase A2, membrane associated, PLA2B, PLA2BPLA2, PLA2L, PLA2LPLA2S, PLAS1, RASF-A, sPLA2 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 5320 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction