Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Plasma Kallikrein/KLKB1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2498SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2498SP 25 μg
MAB2498 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2498SP Supplier R&D Systems Supplier No. MAB2498SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Plasma Kallikrein/KLKB1 Monoclonal specifically detects Plasma Kallikrein/KLKB1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Plasma Kallikrein/KLKB1
Applications Western Blot
Classification Monoclonal
Clone 337801
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_032481
Gene Alias EC 3.4.21, EC 3.4.21.34, Fletcher factor, kallikrein B, plasma (Fletcher factor) 1, kininogenin, KLK3plasma kallikrein, KLKB1, plasma kallikrein heavy chain, plasma kallikrein light chain, Plasma prekallikrein, PPK
Gene Symbols KLKB1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Kallikrein/KLKB1 Gly20-Ala638 Accession # NP_032481
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3818
Test Specificity Detects mouse Plasma Kallikrein/KLKB1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) Kallikrein/KLKB1, rhKallikrein 1, 3, 4, 5, 7, 8, 10, 11, 12, 13, 14, 15, recombinant mouse HGFA, rhFactor VI, rhFactor X, rhFactor XI, rhThrombin, or rhuPA is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.