Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Plasma Kallikrein/KLKB1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2498
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2498 100 μg
AF2498SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2498 Supplier R&D Systems Supplier No. AF2498
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

Plasma Kallikrein/KLKB1 Polyclonal specifically detects Plasma Kallikrein/KLKB1 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Plasma Kallikrein/KLKB1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Gene Accession No. NP_032481
Gene Alias EC 3.4.21, EC 3.4.21.34, Fletcher factor, kallikrein B, plasma (Fletcher factor) 1, kininogenin, KLK3plasma kallikrein, KLKB1, plasma kallikrein heavy chain, plasma kallikrein light chain, Plasma prekallikrein, PPK
Gene Symbols KLKB1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Kallikrein/KLKB1 Gly20-Ala638 Accession # NP_032481
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3818
Test Specificity Detects mouse Plasma Kallikrein/KLKB1 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human KLKB1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.