Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Plasminogen Kringle 5 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF742SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF742SP 25 μg
AF742 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF742SP Supplier R&D Systems Supplier No. AF742SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Plasminogen Kringle 5 Polyclonal specifically detects Plasminogen Kringle 5 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Plasminogen Kringle 5
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAA50168
Gene Symbols Plg
Host Species Goat
Immunogen E. coli-derived recombinant mouse Plasminogen Kringle 5 Cys481-Ala563 Accession # AAA50168
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5340
Test Specificity Detects mouse Plasminogen Kringle 5 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human Angiostatin (Kringle 1-3) is observed. This antibody is expected to show cross-reactivity with mouse plasminogen.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.