Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Plasminogen Kringle 5 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF742
This item is not returnable.
View return policy
Description
Plasminogen Kringle 5 Polyclonal specifically detects Plasminogen Kringle 5 in Mouse samples. It is validated for Western Blot.Specifications
Plasminogen Kringle 5 | |
Polyclonal | |
Biotin | |
AAA50168 | |
Goat | |
Affinity Purified | |
RUO | |
5340 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
Plg | |
E. coli-derived recombinant mouse Plasminogen Kringle 5 Cys481-Ala563 Accession # AAA50168 | |
50 μg | |
Primary | |
Detects mouse Plasminogen Kringle 5 in Western blots. In Western blots, less than 5% cross-reactivity with recombinant human Angiostatin (Kringle 1-3) is observed. This antibody is expected to show cross-reactivity with mouse Plasminogen. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction