Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Plexin B2 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB6836U100UG
This item is not returnable.
View return policy
Description
Plexin B2 Monoclonal specifically detects Plexin B2 in Mouse samples. It is validated for Flow Cytometry.Specifications
Plexin B2 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
B2RXS4 | |
PLXNB2 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse Plexin B2 Leu20-Trp1029 Accession # B2RXS4 | |
RUO | |
23654 | |
Mouse | |
IgG2a |
Flow Cytometry | |
772417 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
MM1, MM1KIAA0315dJ402G11.3, Nbla00445, PLEXB2, plexin B2, plexin-B2, PLXNB2 | |
Rat | |
100 μg | |
Primary | |
Detects mouse Plexin B2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Plexin B2 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction