Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Plexin C1 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF5375
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF5375 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF5375 Supplier R&D Systems Supplier No. BAF5375
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Plexin C1 Polyclonal specifically detects Plexin C1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Plexin C1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q9QZC2
Gene Alias CD232, CD232 antigen, plexin (semaphorin receptor), plexin C1, PLXNC1, PLXN-C1, receptor for viral semaphorin protein, receptor for virally-encoded semaphorin, VESPR, VESPRplexin-C1, Virus-encoded semaphorin protein receptor
Gene Symbols PLXNC1
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Plexin C1 Ala35-Thr950 Accession # Q9QZC2
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10154
Test Specificity Detects mouse Plexin C1 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant mouse Plexin A1, A2, A3, recombinant human Plexin B1, B2, and D1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.