Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse PlGF-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB465R100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB465R100 100 μg
MAB465RSP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB465R100 Supplier R&D Systems Supplier No. MAB465R100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

PlGF-2 Monoclonal specifically detects PlGF-2 in Mouse samples. It is validated for Blockade of Receptor-ligand Interaction.

Specifications

Antigen PlGF-2
Applications Blocking Assay
Classification Monoclonal
Clone 62526R
Conjugate Unconjugated
Dilution Blockade of Receptor-ligand Interaction 0.015-0.05 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P49764
Gene Alias OORS, PlGF2
Host Species Rat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse PlGF-2, Ala24-Pro158, Accession # P49764
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5281
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.