Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse PLTP Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF4918SP
This item is not returnable.
View return policy
Description
PLTP Polyclonal specifically detects PLTP in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PLTP | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
HDLCQ9, Lipid transfer protein II, phospholipid transfer protein | |
Sheep | |
Affinity Purified | |
RUO | |
5360 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
P55065 | |
PLTP | |
Mouse myeloma cell line NS0-derived recombinant mouse PLTP Glu18-Ala493 (Asp32Glu) Accession # P55065 | |
25 μg | |
Primary | |
Detects mouse PLTP in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant human PLTP is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction