Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse PLTP Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF4918
This item is not returnable.
View return policy
Description
PLTP Polyclonal specifically detects PLTP in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PLTP | |
Polyclonal | |
Unconjugated | |
P55065 | |
PLTP | |
Mouse myeloma cell line NS0-derived recombinant mouse PLTP Glu18-Ala493 (Asp32Glu) Accession # P55065 | |
100 μg | |
Primary | |
Detects mouse PLTP in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant human PLTP is observed. | |
Mouse | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
HDLCQ9, Lipid transfer protein II, phospholipid transfer protein | |
Sheep | |
Affinity Purified | |
RUO | |
5360 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction