Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse PLUNC Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF4274
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF4274 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF4274 Supplier R&D Systems Supplier No. BAF4274
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

PLUNC Polyclonal specifically detects PLUNC in Mouse samples. It is validated for Western Blot.

Specifications

Antigen PLUNC
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. P97361
Gene Alias bA49G10.5, BPIFA1, ligand-binding protein RYA3, LPLUNC3, Lung-specific protein X, LUNX, LUNXNASG, Nasopharyngeal carcinoma-related protein, Palate lung and nasal epithelium clone protein, palate, lung and nasal epithelium associated, palate, lung and nasal epithelium carcinoma associated, protein Plunc, Secretory protein in upper respiratory tracts, SPLUNC1, SPLUNC1SPURT, tracheal epithelium enriched protein, Tracheal epithelium-enriched protein, Von Ebner protein Hl
Gene Symbols BPIFA1
Host Species Sheep
Immunogen E. coli-derived recombinant mouse PLUNC Ala59-Val278 Accession # P97361
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51297
Test Specificity Detects mouse PLUNC in Western blots. In Western blots, less than 10% cross-reactivity with recombinant human PLUNC is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.