Learn More
Mouse Podocalyxin Alexa Fluor™ 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody
Supplier:  R&D Systems FAB1556N100UG
Catalog No. FB1556N100U
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
Podocalyxin Like Monoclonal specifically detects Podocalyxin Like in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
Podocalyxin Like
Monoclonal
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with Sodium Azide
GCTM, Gp200MGC138240, PCLP, PCLP1, PCLP-1, PCLPGCTM-2 antigen, PCpodocalyxin, POD XL, podocalyxin-like, Podocalyxin-like protein 1, PODXL
Rat
Protein A or G purified from hybridoma culture supernatant
Primary
Detects mouse Podocalyxin in direct ELISAs and Western blots.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
IgG2b
Flow Cytometry
192703
Flow Cytometry 0.25-1 μg/106 cells
Q9R0M4
PODXL
Mouse myeloma cell line NS0-derived recombinant mouse Podocalyxin, Ser21-Arg402, Accession No. Q9R0M4
100 μg
5420.0
Mouse
Supernatant
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.