Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Podocalyxin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1556
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1556 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB1556 Supplier R&D Systems Supplier No. MAB1556
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Podocalyxin Like Monoclonal specifically detects Podocalyxin Like in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready.

Specifications

Antigen Podocalyxin Like
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 192703
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Flow Cytometry 2.5 μg/10^6 cells, Immunohistochemistry 1-25 μg/mL, Immunocytochemistry 8-25 μg/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9R0M4
Gene Alias GCTM, Gp200MGC138240, PCLP, PCLP1, PCLP-1, PCLPGCTM-2 antigen, PCpodocalyxin, POD XL, podocalyxin-like, Podocalyxin-like protein 1, PODXL
Gene Symbols PODXL
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Podocalyxin Ser21-Arg402 Accession # Q9R0M4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5420
Test Specificity Detects mouse Podocalyxin in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.