Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse PPAR delta (aa 1-14) Synthetic Peptide

Catalog No. PEP026
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP026 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP026 Supplier Invitrogen™ Supplier No. PEP026

Synthetic Peptide

Murine PPAR delta (residues 1-14) Synthetic Peptide, PEP-026, from Invitrogen.

PEP-026 is a 14 amino acid (amino acids 1-14) synthetic peptide whose sequence is derived from murine PPAR delta and is completely unique to PPAR delta. A COOH-terminal cysteine residue was added for conjugation to KLH. The sequence of this peptide is (amino to carboxy terminus): M1-E-Q-P-Q-E-E-T-P-E-A-R-E-E14-CThis peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and PPAR delta, catalog # PA1-823A. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody.Reconstitute with 0.1 ml of distilled water.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 19015
Species Mouse
Purification Method purified
Accession Number P35396
Common Name Mouse PPAR delta (aa 1-14)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol Ppard
Lyophilized Yes
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence M1 - E - Q - P - Q - E - E - T - P - E - A - R - E - E14 - C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.