Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse PRAT4A Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF4429
This item is not returnable.
View return policy
Description
PRAT4A/TNRC5 Polyclonal specifically detects PRAT4A/TNRC5 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PRAT4A/TNRC5 | |
Polyclonal | |
Unconjugated | |
Q9DAU1 | |
CNPY3 | |
Mouse myeloma cell line NS0-derived recombinant mouse PRAT4A isoform 1 Glu38-Leu276 Accession # Q9DAU1 | |
100 μg | |
Primary | |
Detects mouse PRAT4A in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse PRAT4B is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
CAG4A, canopy 3 homolog (zebrafish), CNPY3, CTG4A, ERDA5, ERDA5CTG repeat protein 4a, Expanded repeat-domain protein CAG/CTG 5, PRAT4ACAG repeat containing, Protein associated with TLR4, protein associated with Toll-like receptor 4A, protein canopy homolog 3, TNRC5, TNRC5expanded repeat domain, CAG/CTG 5, trinucleotide repeat containing 5, Trinucleotide repeat-containing gene 5 protein | |
Goat | |
Affinity Purified | |
RUO | |
10695 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction