Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse PRDC/GREM2 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | PRDC/GREM2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Description
PRDC/GREM2 Polyclonal specifically detects PRDC/GREM2 in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
PRDC/GREM2 | |
Western Blot 0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
CKTSF1B2, CKTSF1B2FLJ21195, Cysteine knot superfamily 1, BMP antagonist 2, DAN domain family member 3, DAND3, DAND3Prdc, GREM2, gremlin 2, gremlin 2, cysteine knot superfamily, homolog, gremlin-2, PRDCgremlin 2, cysteine knot superfamily, homolog (Xenopus laevis), Protein related to DAN and cerberus | |
GREM2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, ELISA | |
Unconjugated | |
RUO | |
O88273 | |
64388 | |
E. coli-derived recombinant mouse PRDC Arg22-Gln168 Accession # O88273 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse PRDC in ELISAs and Western blots. In sandwich ELISAs, less than 0.1% cross-reactivity with recombinant mouse (rm) Cerberus, rmGremlin, rmDAN, recombinant human (rh) COCO, rhGremlin, and rmBMP-4 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title