Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse PRDC/GREM2 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF2069
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF2069 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF2069 Supplier R&D Systems Supplier No. BAF2069
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

PRDC/GREM2 Polyclonal specifically detects PRDC/GREM2 in Mouse samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).

Specifications

Antigen PRDC/GREM2
Applications Western Blot, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL
Gene Accession No. O88273
Gene Alias CKTSF1B2, CKTSF1B2FLJ21195, Cysteine knot superfamily 1, BMP antagonist 2, DAN domain family member 3, DAND3, DAND3Prdc, GREM2, gremlin 2, gremlin 2, cysteine knot superfamily, homolog, gremlin-2, PRDCgremlin 2, cysteine knot superfamily, homolog (Xenopus laevis), Protein related to DAN and cerberus
Gene Symbols GREM2
Host Species Goat
Immunogen E. coli-derived recombinant mouse PRDC/GREM2 Arg22-Gln168 Accession # O88273
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64388
Test Specificity Detects mouse PRDC/GREM2 in ELISAs and Western blots. In sandwich immunoassays, less than 0.1% cross-reactivity with recombinant mouse (rm) Cerberus, rmGremlin, rmDAN, recombinant human (rh) COCO, rhGremlin, and rmBMP-4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.