Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Pref-1/DLK1/FA1 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Catalog No. FAB8634G025
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB8634G025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB8634G025 Supplier R&D Systems Supplier No. FAB8634G025
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Pref-1/DLK1/FA1 Monoclonal specifically detects Pref-1/DLK1/FA1 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Pref-1/DLK1/FA1
Applications Flow Cytometry
Classification Monoclonal
Clone 1168B
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q09163
Gene Alias delta-like 1 homolog (Drosophila), DLK, DLK1, DLK-1, FA1, FA1fetal antigen 1, pG2, pG2delta-like homolog (Drosophila), preadipocyte factor 1, Pref-1, PREF1, protein delta homolog 1, secredeltin, ZOG
Gene Symbols DLK1
Host Species Rabbit
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Pref-1/DLK1/FA1 Ala24-Gln305 Accession # Q09163
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8788
Test Specificity Detects mouse Pref-1 in direct ELISAs.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.