Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Pref-1/DLK1/FA1 Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB8634R100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB8634R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB8634R100 Supplier R&D Systems Supplier No. FAB8634R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Pref-1/DLK1/FA1 Monoclonal specifically detects Pref-1/DLK1/FA1 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Pref-1/DLK1/FA1
Applications Flow Cytometry
Classification Monoclonal
Clone 1168B
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q09163
Gene Alias delta-like 1 homolog (Drosophila), DLK, DLK1, DLK-1, FA1, FA1fetal antigen 1, pG2, pG2delta-like homolog (Drosophila), preadipocyte factor 1, Pref-1, PREF1, protein delta homolog 1, secredeltin, ZOG
Host Species Rabbit
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Pref-1/DLK1/FA1 Ala24-Gln305 Accession # Q09163
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8788
Test Specificity Detects mouse Pref-1 in direct ELISAs.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.